Learn More
Medchemexpress LLC Mycophenolate mofetil | 128794-94-5 | 99.8% | 200 MG
SDP
Catalog No. 5000407336
Encompass_Preferred
Mycophenolate mofetil (RS 61443) is the morpholinoethylester proagent of Mycophenolic acid. It inhibits de novo purine synthesis by inhibiting inosine monophosphate dehydrogenase (IMPDH). Mycophenolate mofetil exhibits selective lymphocyte antiproliferative effects on both T and B cells, which prevents antibody formation.
Catalog No. 50-004-07336 Supplier Medchemexpress LLC Supplier No. HYB0199200MG
Mycophenolate mofetil (RS 61443) is the morpholinoethylester proagent of Mycophenolic acid. It inhibits de novo purine synthesis by inhibiting inosine monophosphate dehydrogenase (IMPDH). Mycophenolate mofetil exhibits selective lymphocyte antiproliferative effects on both T and B cells, which prevents antibody formation.