Learn More
Enzo Life Sciences Mycophenolate mofetIL-(20mg). CAS: 128794-94-5
SDP
Catalog No. 502011043
Encompass_Preferred
Mycophenolate mofetil (CellCept) is an immunosuppressant prodrug of mycophenolic acid, a fermentation product of Penicillium brevicompactum. Immunosuppression occurs via inhibition of type II inosine-5’-monophosphate dehydrogenase, depleting guanosine preferentially in T and B lymphocytes. In addition, mycophenolic acid displays anti-inflammatory activity by preventing NO production specifically from iNOS and by blocking glycosylation and expression of adhesion molecules and therefore recruitment of lymphocytes and monocytes to sites of inflammation. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml, warm). Long Term Storage: Ambient.
Catalog No. 50-201-1043 Supplier Enzo Life Sciences Supplier No. BMLIM1010020
Mycophenolate mofetil (CellCept) is an immunosuppressant prodrug of mycophenolic acid, a fermentation product of Penicillium brevicompactum. Immunosuppression occurs via inhibition of type II inosine-5’-monophosphate dehydrogenase, depleting guanosine preferentially in T and B lymphocytes. In addition, mycophenolic acid displays anti-inflammatory activity by preventing NO production specifically from iNOS and by blocking glycosylation and expression of adhesion molecules and therefore recruitment of lymphocytes and monocytes to sites of inflammation. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml, warm). Long Term Storage: Ambient.