Learn More
Apexbio Technology LLC Mycophenolate Mofetil 128794-94-5 50mg
SDP
Catalog No. 502240568
Encompass_Preferred
Mycophenolate Mofetil (128794-94-5) is a small-molecule inhibitor targeting inosine monophosphate dehydrogenase (IMPDH) isoforms I and II It is designed to selectively and reversibly inhibit IMPDH thereby disrupting de novo guanosine nucleotide biosynthesis Mycophenolate Mofetil exerts its biological activity primarily through non-competitive inhibition of IMPDH blocking the conversion of inosine monophosphate (IMP) to xanthosine monophosphate (XMP) In in vitro studies Mycophenolate Mofetil demonstrates inhibitory activity with IC50 values of 39 nM and 27 nM for IMPDH I and II respectively Based on these pharmacological properties Mycophenolate Mofetil holds research potential in studies on immunology purine metabolism regulation cell proliferation transplant rejection and autoimmune disease models
This item is not returnable. View return policy
Catalog No. 50-224-0568 Supplier Apexbio Technology LLC Supplier No. A433650
Mycophenolate Mofetil (128794-94-5) is a small-molecule inhibitor targeting inosine monophosphate dehydrogenase (IMPDH) isoforms I and II It is designed to selectively and reversibly inhibit IMPDH thereby disrupting de novo guanosine nucleotide biosynthesis Mycophenolate Mofetil exerts its biological activity primarily through non-competitive inhibition of IMPDH blocking the conversion of inosine monophosphate (IMP) to xanthosine monophosphate (XMP) In in vitro studies Mycophenolate Mofetil demonstrates inhibitory activity with IC50 values of 39 nM and 27 nM for IMPDH I and II respectively Based on these pharmacological properties Mycophenolate Mofetil holds research potential in studies on immunology purine metabolism regulation cell proliferation transplant rejection and autoimmune disease models