Learn More
Cayman Chemical Mycophenolate Mofetil-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1132748-21-0, 1 mg
SDP
Catalog No. 502932531
Encompass_Preferred
An internal standard for the quantification of mycophenolate mofetil by GC- or LC-MS
Catalog No. 50-293-2531 Supplier Cayman Chemical Supplier No. 252811 MG
An internal standard for the quantification of mycophenolate mofetil by GC- or LC-MS