Learn More
Cayman Chemical Mycophenolate MofetIl-d4 500ug
SDP
Catalog No. 502932524
Encompass_Preferred
An internal standard for the quantification of mycophenolate mofetil by GC- or LC-MS
Catalog No. 50-293-2524 Supplier Cayman Chemical Supplier No. 25281500 UG
An internal standard for the quantification of mycophenolate mofetil by GC- or LC-MS