Learn More
Apexbio Technology LLC Mycophenolate mofetil hydrochloride 116680-01-4 200mg
SDP
Catalog No. 5000088355
Encompass_Preferred
Mycophenolate mofetil hydrochloride (CAS 116680-01-4) is a small-molecule inhibitor targeting inosine monophosphate dehydrogenase (IMPDH) types I and II It is designed to selectively and reversibly inhibit IMPDH thereby modulating the de novo biosynthesis pathway of purine nucleotides Mycophenolate mofetil hydrochloride exerts its biological activity primarily through inhibition of IMPDH It demonstrates inhibitory activity with IC50 values of 39 nM for IMPDH type I and 27 nM for IMPDH type II Based on these pharmacological properties mycophenolate mofetil hydrochloride holds research potential in the study of immune processes organ transplant rejection mechanisms and autoimmune disease models
Catalog No. 50-000-88355 Supplier Apexbio Technology LLC Supplier No. B1101200
Mycophenolate mofetil hydrochloride (CAS 116680-01-4) is a small-molecule inhibitor targeting inosine monophosphate dehydrogenase (IMPDH) types I and II It is designed to selectively and reversibly inhibit IMPDH thereby modulating the de novo biosynthesis pathway of purine nucleotides Mycophenolate mofetil hydrochloride exerts its biological activity primarily through inhibition of IMPDH It demonstrates inhibitory activity with IC50 values of 39 nM for IMPDH type I and 27 nM for IMPDH type II Based on these pharmacological properties mycophenolate mofetil hydrochloride holds research potential in the study of immune processes organ transplant rejection mechanisms and autoimmune disease models