Learn More
Abcam Mycophenolate mofetil, MPDH inhibitor, 50MG
Catalog No. 502706138
Encompass_Preferred
MW 433.5 Da, Purity >98%. Selective, non-competitive and reversible IMPDH inhibitor (IC₅₀ = 3 μM). Immunosuppressant agent. Inhibits guanosine synthesis. Shows antiparasitic effects and suppresses T cells and B cells in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6138 Supplier Abcam Supplier No. AB14301350MG
MW 433.5 Da, Purity >98%. Selective, non-competitive and reversible IMPDH inhibitor (IC₅₀ = 3 μM). Immunosuppressant agent. Inhibits guanosine synthesis. Shows antiparasitic effects and suppresses T cells and B cells in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement