Learn More
Cayman Chemical Mycophenolic Acid methyl ester, Purity: ≥98%, CAS: 31858-66-9, 25 mg
SDP
Catalog No. 502917841
Encompass_Preferred
An esterified form of mycophenolic acid; inhibits pioglitazone-induced transactivation of PPARγ in U2OS cells in a reporter assay at 1 and 10 µM; cytotoxic against a panel of seven cancer cell lines (IC50s = 0.26-23.73 µM); inhibits tumor growth in an Ehrlich murine spontaneous adenocarcinoma model and decreases the production of antibodies against sheep red blood cells in mice at 120 mg/kg; has been used as a precursor in the synthesis of mycophenolic acid β-D-glucuronide; a potential impurity in commercial preparations of mycophenolate mofetil
Catalog No. 50-291-7841 Supplier Cayman Chemical Supplier No. 3839725 MG
An esterified form of mycophenolic acid; inhibits pioglitazone-induced transactivation of PPARγ in U2OS cells in a reporter assay at 1 and 10 µM; cytotoxic against a panel of seven cancer cell lines (IC50s = 0.26-23.73 µM); inhibits tumor growth in an Ehrlich murine spontaneous adenocarcinoma model and decreases the production of antibodies against sheep red blood cells in mice at 120 mg/kg; has been used as a precursor in the synthesis of mycophenolic acid β-D-glucuronide; a potential impurity in commercial preparations of mycophenolate mofetil