Learn More
Medchemexpress LLC Mycophenolic acid | 24280-93-1 | 100 MG
SDP
Catalog No. 501874098
Encompass_Preferred
Mycophenolic acid is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with an EC50 of 0.24 μM. It demonstrates antiviral effects against a wide range of RNA viruses, including influenza, dengue virus, Zika virus, rotavirus, CCHFV, and hantavirus. This immunosuppressive agent also exhibits antiangiogenic and antitumor effects.
  • Potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor.
  • Demonstrates antiviral effects against various RNA viruses.
  • Acts as an immunosuppressive agent.
  • Exhibits antiangiogenic and antitumor effects.
  • Shows preferential antiproliferative activity against endothelial cells and fibroblasts.
  • Can down-regulate HDAC2 and MYC, and up-regulate NDRG1.
Catalog No. 50-187-4098 Supplier Medchemexpress LLC Supplier No. HYB0421100MG
Mycophenolic acid is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with an EC50 of 0.24 μM. It demonstrates antiviral effects against a wide range of RNA viruses, including influenza, dengue virus, Zika virus, rotavirus, CCHFV, and hantavirus. This immunosuppressive agent also exhibits antiangiogenic and antitumor effects.
  • Potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor.
  • Demonstrates antiviral effects against various RNA viruses.
  • Acts as an immunosuppressive agent.
  • Exhibits antiangiogenic and antitumor effects.
  • Shows preferential antiproliferative activity against endothelial cells and fibroblasts.
  • Can down-regulate HDAC2 and MYC, and up-regulate NDRG1.