Learn More
Alkali Scientific Mycophenolic Acid, 250 Milligrams
SDP
Catalog No. 5000661328
Encompass_Preferred
Mycophenolic Acid is an immunosuppressant compound used primarily in cell culture applications. It is a highly pure compound commonly used in biochemical research and therapeutic studies. As an antibiotic in its class, Mycophenolic Acid inhibits the proliferation of certain immune cells by interfering with the de novo purine biosynthesis pathway. This makes it useful for studying immune responses and cellular functions in vitro. It is also used in various molecular biology applications, especially for investigating immune modulation. Available in a 250-milligram quantity, it is suitable for experiments requiring high purity for optimal results in cell culture environments.
Catalog No. 50-006-61328 Supplier Alkali Scientific Supplier No. MYC002CP
Mycophenolic Acid is an immunosuppressant compound used primarily in cell culture applications. It is a highly pure compound commonly used in biochemical research and therapeutic studies. As an antibiotic in its class, Mycophenolic Acid inhibits the proliferation of certain immune cells by interfering with the de novo purine biosynthesis pathway. This makes it useful for studying immune responses and cellular functions in vitro. It is also used in various molecular biology applications, especially for investigating immune modulation. Available in a 250-milligram quantity, it is suitable for experiments requiring high purity for optimal results in cell culture environments.