Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycophenolic acid, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019065 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9065 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9065 Supplier MilliporeSigma Supelco Supplier No. 8928710MG
Add to Cart
Add to Cart

Mycophenolic acid is an active metabolite of the ester prodrug mycophenolate mofetil (MPM), which is prescribed as an immunosuppressant to prevent the organ transplantation rejection and also in the treatment of certain autoimmune diseases.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 24280-93-1
Physical Form Neat
Linear Formula C17H20O6
MDL Number MFCD00036814
Synonym 6-(1,3-Dihydro-7-hydroxy-5-methoxy-4-methyl-1-oxoisobenzofuran-6-yl)-4-methyl-4-hexanoic acid; 6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl)-4-methyl-4-hexenoic acid; NSC 129185
Type Analytical Standard
Quantity 10 mg
Formula Weight 320.34
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.