Learn More
TARGETMOL CHEMICALS INC Mycophenolic acid-beta-D-glucu
SDP
Catalog No. 502880125
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Mycophenolic acid glucuronide is the glucuronide metabolite of mycophenolic acid in human plasma. Purity 99%
Catalog No. 50-288-0125 Supplier TARGETMOL CHEMICALS INC Supplier No. T601045MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Mycophenolic acid glucuronide is the glucuronide metabolite of mycophenolic acid in human plasma. Purity 99%