Learn More
Medchemexpress LLC HY-B0421S 1mg Medchemexpress, Mycophenolic acid D3 CAS:1185242-90-3 Purity:>98%
SDP
Catalog No. 501932914
Encompass_Preferred
Medchemexpress, HY-B0421S 1mg Mycophenolic acid D3 CAS:1185242-90-3 Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2914 Supplier Medchemexpress LLC Supplier No. HYB0421S1MG
Medchemexpress, HY-B0421S 1mg Mycophenolic acid D3 CAS:1185242-90-3 Mycophenolic acid D3 (Mycophenolate D3) is deuterium labeled Mycophenolic acid, which is an an immunosuppresant drug and has potent anti-proliferative activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.