Learn More
Cayman Chemical Mycophenolic Acid β-D-Glucuronide, Purity: ≥98%, CAS: 31528-44-6, 5 mg
SDP
Catalog No. 502981806
Encompass_Preferred
A metabolite of mycophenolic acid; does not inhibit IMPDH type II in a cell-free assay; reduces tumor growth in an Ehrlich murine spontaneous adenocarcinoma model at 6 mg/animal
Catalog No. 50-298-1806 Supplier Cayman Chemical Supplier No. 190785 MG
A metabolite of mycophenolic acid; does not inhibit IMPDH type II in a cell-free assay; reduces tumor growth in an Ehrlich murine spontaneous adenocarcinoma model at 6 mg/animal