Learn More
Medchemexpress LLC Mycophenolic acid glucuronide | 31528-44-6 | MFCD00241088 | 99.6% | 496.46 | 1 ML
SDP
Catalog No. 5000349626
Encompass_Preferred
Mycophenolic acid glucuronide is a metabolite of the immunosuppressant mycophenolic acid (MPA).
  • Shows anti-tumor activity.
  • Can be used in adenocarcinoma research.
  • Shows weak IMP dehydrogenase (IMPDH) type II inhibition with an IC50 of 3.3 mg/L.
  • Reduces tumor growth in an Ehrlich murine spontaneous adenocarcinoma model with 76.8% tumor inhibition compared to the untreated control.
Catalog No. 50-003-49626 Supplier Medchemexpress LLC Supplier No. HY13730110MM/1ML
Add to Cart
Add to Cart
Mycophenolic acid glucuronide is a metabolite of the immunosuppressant mycophenolic acid (MPA).
  • Shows anti-tumor activity.
  • Can be used in adenocarcinoma research.
  • Shows weak IMP dehydrogenase (IMPDH) type II inhibition with an IC50 of 3.3 mg/L.
  • Reduces tumor growth in an Ehrlich murine spontaneous adenocarcinoma model with 76.8% tumor inhibition compared to the untreated control.