Learn More
Medchemexpress LLC Mycophenolic acid (Standard) | 24280-93-1 | 100.0% | 50 MG
SDP
Catalog No. 5000409248
Encompass_Preferred
Mycophenolic acid (Standard) is the analytical standard of Mycophenolic acid, intended for research and analytical applications. It is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with an EC50 of 0.24 μM, demonstrating antiviral effects against various RNA viruses, including influenza. It also acts as an immunosuppressive agent with antiangiogenic and antitumor effects. This compound is used as a reference standard for assay in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS. It is for research use only and not sold to patients.
  • Potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor
  • Exhibits antiviral effects against various RNA viruses
  • Acts as an immunosuppressive agent
  • Demonstrates antiangiogenic and antitumor effects
  • Used in qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-09248 Supplier Medchemexpress LLC Supplier No. HYB0421R50MG
Add to Cart
Add to Cart
Mycophenolic acid (Standard) is the analytical standard of Mycophenolic acid, intended for research and analytical applications. It is a potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor with an EC50 of 0.24 μM, demonstrating antiviral effects against various RNA viruses, including influenza. It also acts as an immunosuppressive agent with antiangiogenic and antitumor effects. This compound is used as a reference standard for assay in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS. It is for research use only and not sold to patients.
  • Potent uncompetitive inosine monophosphate dehydrogenase (IMPDH) inhibitor
  • Exhibits antiviral effects against various RNA viruses
  • Acts as an immunosuppressive agent
  • Demonstrates antiangiogenic and antitumor effects
  • Used in qualitative, quantitative, and methodological research experiments