Learn More
HIMEDIA Mycoplasma Agar Base (PPLO Agar Base), M266, Microbiology Culture Media, Dehydrated Culture Media, Agar Base, Mycoplasma Isolation, Requires Enrichment, Powder, 500 g
Catalog No. NC4013732
Encompass_Preferred
Mycoplasma Agar Base, also known as PPLO Agar Base, supports the isolation and cultivation of Mycoplasma species (pleuropneumonia-like organisms) when supplemented with serum and yeast enrichment. A generous heart infusion and peptone base with sodium chloride and agar, final pH 7.8, provides the sterols and growth factors these wall-less, highly fastidious organisms demand. Widely used in veterinary, cell culture contamination and clinical mycoplasmology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013732 Supplier HIMEDIA Supplier No. M266500G
Mycoplasma Agar Base, also known as PPLO Agar Base, supports the isolation and cultivation of Mycoplasma species (pleuropneumonia-like organisms) when supplemented with serum and yeast enrichment. A generous heart infusion and peptone base with sodium chloride and agar, final pH 7.8, provides the sterols and growth factors these wall-less, highly fastidious organisms demand. Widely used in veterinary, cell culture contamination and clinical mycoplasmology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.