Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mycoplasma Arginine Deiminase Recombinant Protein, PeproTech®

Catalog No. p-7246368
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
This item is not returnable. View return policy

Catalog No. 15012100UG

Supplier: Gibco™ 15012100UG

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Mycoplasma Arginine Deiminase is a 46.3 kDa protein consisting of 409 amino acids. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Arginine Deiminase (ADI) is a microbial enzyme from Mycoplasma produced in E.coli. It has high affinity to L-arginine and hydrolyzes L-arginine to citrulline and ammonia. Low concentrations of ADI have been shown to inhibit proliferation in certain cultured cells by arresting the cell cycle in G1 and/or S phase. Higher concentrations of ADI lead to subsequent apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P0A9Q1, P23793
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 69703137, 82262143
Molecular Weight (g/mol) 46.3 kDa
Name Mycoplasma Arginine Deiminase
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name Arginine Deiminase
Gene Symbol arcA
Biological Activity Measured by its ability to induce apoptosis in Jurkat cells using a concentration of 100-150 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence SVFDSKFKGI HVYSEIGELE SVLVHEPGRE IDYITPARLD ELLFSAILES HDARKEHKQF VAELKANDIN VVELIDLVAE TYDLASQEAK DKLIEEFLED SEPVLSEEHK VVVRNFLKAK KTSRELVEIM MAGITKTDLG IEADHELIVD PMPNLYFTRD PFASVGNGVT IHYMRYKVRQ RETLFSRFVF SNHPKLINTP WYYDPSLKLS IEGGDVFIYN NDTLVVGVSE RTDLQTVTLL AKNIVANKEC EFKRIVAINV PKWTNLMHLD TWLTMLDKDK FLYSPIANDV FKFWDYDLVN GGAEPQPVEN GLPLEGLLQS IINKKPVLIP IAGEGASQME IERETHFDGT NYLAIRPGVV IGYSRNEKTN AALEAAGIKV LPFHGNQLSL GMGNARCMSM PLSRKDVKW
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.