Learn More
ATCC Mycoplasma arginini; Quantitative CRM; genomic DNA
Catalog No. 502384018
Encompass
Quantified genomic DNA isolated from Mycoplasma arginini strain G230 [NCTC 10129] (ATCC 23838). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-4018 Supplier ATCC Supplier No. QCRM23838D
 Restricted Item
Add to Cart
Add to Cart
Quantified genomic DNA isolated from Mycoplasma arginini strain G230 [NCTC 10129] (ATCC 23838). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.