Learn More
TRIBIOSCIENCE INC Mycoplasma Detection qPCR Kit, 100 Reactions
SDP
Catalog No. NC2158181
Encompass
Mycoplasma Species qPCR Kit has been designed to specifically identify the mycoplasma genus species in a single PCR reaction using real-time quantitative polymerase chain reaction (qPCR) and probe fluorescence labels. The kit provides a validated DNA preparation and qPCR procedures for mycoplasma species detection.
This item is not returnable. View return policy
Catalog No. NC2158181 Supplier TRIBIOSCIENCE INC Supplier No. TBS42030100
Mycoplasma Species qPCR Kit has been designed to specifically identify the mycoplasma genus species in a single PCR reaction using real-time quantitative polymerase chain reaction (qPCR) and probe fluorescence labels. The kit provides a validated DNA preparation and qPCR procedures for mycoplasma species detection.