Learn More
TRIBIOSCIENCE INC Mycoplasma Detection qPCR Kit, 200 Reactions
SDP
Catalog No. NC2944533
Encompass
Mycoplasma Species qPCR Kit has been designed to specifically identify the mycoplasma genus species in a single PCR reaction using real-time quantitative polymerase chain reaction (qPCR) and probe fluorescence labels. The kit provides a validated DNA preparation and qPCR procedures for mycoplasma species detection.
Catalog No. NC2944533 Supplier TRIBIOSCIENCE INC Supplier No. TBS42030200
Mycoplasma Species qPCR Kit has been designed to specifically identify the mycoplasma genus species in a single PCR reaction using real-time quantitative polymerase chain reaction (qPCR) and probe fluorescence labels. The kit provides a validated DNA preparation and qPCR procedures for mycoplasma species detection.