Learn More
TRIBIOSCIENCE INC Mycoplasma Eraser Solution, 2 ML
SDP
Catalog No. NC3454601
Encompass_Preferred
Tribioscience’s Mycoplasma Eraser is designed to clean up mycoplasma contamination from cultured cells when it has been added into culture medium. The activity of solution is stable in culture medium and not affected by penicillin, streptomycin, and serum.
Catalog No. NC3454601 Supplier TRIBIOSCIENCE INC Supplier No. TBS8006
Tribioscience’s Mycoplasma Eraser is designed to clean up mycoplasma contamination from cultured cells when it has been added into culture medium. The activity of solution is stable in culture medium and not affected by penicillin, streptomycin, and serum.