Learn More
ATCC Mycoplasma fermentans; PG18
Catalog No. 502383530
Encompass
Mycoplasma fermentans strain PG18 [G, NCTC 10117] is a bacterial type strain that was isolated from a case of ulcerative balanitis. This product is titered and can be used in assay development and quality control.
Catalog No. 50-238-3530 Supplier ATCC Supplier No. 19989TTR
Mycoplasma fermentans strain PG18 [G, NCTC 10117] is a bacterial type strain that was isolated from a case of ulcerative balanitis. This product is titered and can be used in assay development and quality control.