Learn More
ATCC Mycoplasma fermentans; PG18
Supplier:  ATCC 19989
Encompass
Mycoplasma fermentans strain PG18 [G, NCTC 10117] is a whole-genome sequenced bacterial type strain and that isolated from a case of ulcerative balanitis. This product has applications in quality control and pharmaceutical and personal care research.
Catalog No. 50-238-3408
 Restricted Item