Learn More
ATCC Genomic DNA from Mycoplasma fermentans strain PG18 (ATCC qCRM-19989D™)
Catalog No. 502382790
Encompass
Genomic DNA from Mycoplasma fermentans strain PG18 (ATCC qCRM-19989D™) is a quantified certified reference material that supports USP General Chapter <63>. Produced under an ISO 17034-accredited process, it is used for inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-2790 Supplier ATCC Supplier No. QCRM19989D
 Restricted Item
Add to Cart
Add to Cart
Genomic DNA from Mycoplasma fermentans strain PG18 (ATCC qCRM-19989D™) is a quantified certified reference material that supports USP General Chapter <63>. Produced under an ISO 17034-accredited process, it is used for inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.