Learn More
ATCC Mycoplasma gallisepticum
Catalog No. 502381911
Encompass
Mycoplasma gallisepticum strain [NCTC 10115, PG 31, X95] is bacterial type strain that was isolated from a suspension of tracheal and airsac tissues from chickens with chronic respiratory disease. This product has applications in media testing, quality control, and agricultural research.
Catalog No. 50-238-1911 Supplier ATCC Supplier No. 19610
Mycoplasma gallisepticum strain [NCTC 10115, PG 31, X95] is bacterial type strain that was isolated from a suspension of tracheal and airsac tissues from chickens with chronic respiratory disease. This product has applications in media testing, quality control, and agricultural research.