Learn More
ATCC Genomic DNA from Mycoplasma gallisepticum (ATCC qCRM-19610D™)
Catalog No. 502382174
Encompass
Genomic DNA from Mycoplasma gallisepticum (ATCC qCRM-19610D™) is a quantified certified reference material that supports USP General Chapter <63>. Produced under an ISO 17034-accredited process, it is used for inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-2174 Supplier ATCC Supplier No. QCRM19610D
Genomic DNA from Mycoplasma gallisepticum (ATCC qCRM-19610D™) is a quantified certified reference material that supports USP General Chapter <63>. Produced under an ISO 17034-accredited process, it is used for inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.