Learn More
ATCC Mycoplasma genitalium strain G37 (ATCC 33530™)
Catalog No. 502382108
Encompass
Mycoplasma genitalium strain G37 (ATCC 33530™) is a whole-genome sequenced type strain isolated from the urethra of a male patient with non-gonococcal urethritis. This bacterial culture has applications in infectious disease and sexually transmitted disease research. This culture produces a surface attachment protein.
Catalog No. 50-238-2108 Supplier ATCC Supplier No. 33530
Mycoplasma genitalium strain G37 (ATCC 33530™) is a whole-genome sequenced type strain isolated from the urethra of a male patient with non-gonococcal urethritis. This bacterial culture has applications in infectious disease and sexually transmitted disease research. This culture produces a surface attachment protein.