Learn More
Drg International Inc Mycoplasma Hominis Igg Elisa
Catalog No. 501947612
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Mycoplasma hominis in serum and plasma. The DRG Mycoplasma hominis IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Mycoplasma hominis surface antigen p120. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Mycoplasma hominis-specific antibodies of positive specimens and controls are bound to the immobilized antigens.
This item is not returnable. View return policy
Catalog No. 50-194-7612 Supplier Drg International Inc Supplier No. EIA4559
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Mycoplasma hominis in serum and plasma. The DRG Mycoplasma hominis IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Mycoplasma hominis surface antigen p120. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Mycoplasma hominis-specific antibodies of positive specimens and controls are bound to the immobilized antigens.