Learn More
ATCC Mycoplasma hyorhinis; BTS-7
Catalog No. 502382542
Encompass
Mycoplasma hyorhinis strain BTS-7 is a whole-genome sequenced bacterial type strain that was isolated from the nasal cavity of a pig. It has applications as a control strain, and it is referenced in USP <63>, EP 2.6.7, and JP 16 G3 as a Mycoplasma test organism.
For every order of this item, you must provide a valid Permit for Importation and Transportation of Controlled Materials and Organisms and Vectors (VS Form 16-6A) obtained from the United States Department of Agriculture (USDA), Animal and Plant Health Inspection Service. This item also requires a BSL 2 upgrade before purchase. This form can be found at www.atcc.org
Catalog No. 50-238-2542 Supplier ATCC Supplier No. 17981
Mycoplasma hyorhinis strain BTS-7 is a whole-genome sequenced bacterial type strain that was isolated from the nasal cavity of a pig. It has applications as a control strain, and it is referenced in USP <63>, EP 2.6.7, and JP 16 G3 as a Mycoplasma test organism.
For every order of this item, you must provide a valid Permit for Importation and Transportation of Controlled Materials and Organisms and Vectors (VS Form 16-6A) obtained from the United States Department of Agriculture (USDA), Animal and Plant Health Inspection Service. This item also requires a BSL 2 upgrade before purchase. This form can be found at www.atcc.org