Learn More
ATCC Mycoplasma hyorhinis; control DNA
Supplier:  ATCC 17981D
Encompass
Genomic DNA isolated from Mycoplasma hyorhinis strain BTS-7 (ATCC 17981). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2559