Learn More
ATCC Mycoplasma hyorhinis; Strain BTS-7
Catalog No. 502383571
Encompass
Mycoplasma hyorhinis strain BTS-7 is a whole genome-sequenced strain and type strain isolated from the nasal cavity of a pig. It has applications as a control strain, and is referenced in USP <63>, EP 2.6.7, and JP 16 G3 as a Mycoplasma test organism.
Catalog No. 50-238-3571 Supplier ATCC Supplier No. 17981TTR
Add to Cart
Add to Cart
Mycoplasma hyorhinis strain BTS-7 is a whole genome-sequenced strain and type strain isolated from the nasal cavity of a pig. It has applications as a control strain, and is referenced in USP <63>, EP 2.6.7, and JP 16 G3 as a Mycoplasma test organism.