Learn More
Apexbio Technology LLC MYCOPLASMA LAMP HNB KIT-50mg
SDP
Catalog No. 5000897425
Encompass_Preferred
Mycoplasma LAMP Detection Kit (HNB) is a specialized tool used in the field of biomedical research designed for the rapid detection of mycoplasma contamination in cell cultures Originating from nucleic acid amplification technology this kit utilizes loop-mediated isothermal amplification (LAMP) with a colorimetric indicator hydroxynaphthol blue (HNB) to identify the presence of mycoplasma DNA The mechanism of action targets the conserved sequences in the mycoplasma genome enabling precise and reliable identification This kit exhibits high sensitivity and specificity with detection capabilities in the nanomolar range It is commonly applied in in vitro settings aiding researchers in detecting contamination in cell lines validating cell culture purity and ensuring the accuracy of experimental results Applications extend to drug screening where maintaining uncontaminated cultures is critical Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97425 Supplier Apexbio Technology LLC Supplier No. K282750
Mycoplasma LAMP Detection Kit (HNB) is a specialized tool used in the field of biomedical research designed for the rapid detection of mycoplasma contamination in cell cultures Originating from nucleic acid amplification technology this kit utilizes loop-mediated isothermal amplification (LAMP) with a colorimetric indicator hydroxynaphthol blue (HNB) to identify the presence of mycoplasma DNA The mechanism of action targets the conserved sequences in the mycoplasma genome enabling precise and reliable identification This kit exhibits high sensitivity and specificity with detection capabilities in the nanomolar range It is commonly applied in in vitro settings aiding researchers in detecting contamination in cell lines validating cell culture purity and ensuring the accuracy of experimental results Applications extend to drug screening where maintaining uncontaminated cultures is critical Experimental concentrations typically depend on the specific research design