Learn More
ATCC Mycoplasma orale strain CH 19299 (ATCC 23714™)
Catalog No. 502383039
Encompass
Mycoplasma orale strain CH 19299 (ATCC 23714™) is a whole-genome sequenced type strain isolated from the oropharynx of a human child in Washington, DC. This bacterium has applications in media testing and pharmaceutical QC, and it is recommended as a test organism for USP <63> mycoplasma testing.
Catalog No. 50-238-3039 Supplier ATCC Supplier No. 23714
Mycoplasma orale strain CH 19299 (ATCC 23714™) is a whole-genome sequenced type strain isolated from the oropharynx of a human child in Washington, DC. This bacterium has applications in media testing and pharmaceutical QC, and it is recommended as a test organism for USP <63> mycoplasma testing.