Learn More
ATCC Mycoplasma orale; Quantitative CRM; genomic DNA
Catalog No. 502384631
Encompass
Quantified genomic DNA isolated from Mycoplasma orale strain CH 19299 [NCTC 10112] (ATCC 23714). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-4631 Supplier ATCC Supplier No. QCRM23714D
Quantified genomic DNA isolated from Mycoplasma orale strain CH 19299 [NCTC 10112] (ATCC 23714). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.