Learn More
ATCC Mycoplasma pneumoniae (ATCC 15531™)
Supplier:  ATCC 15531
Encompass
Mycoplasma pneumoniae FH strain of Eaton Agent (ATCC 15531™) is a whole-genome sequenced strain. It is the first mycoplasma identified as a human pathogen and is widely used in respiratory disease research, media testing, and pharmaceutical QC. This strain is recommended as a test organism for USP <63> mycoplasma testing.
Catalog No. 50-238-1678