Learn More
ATCC Mycoplasma pneumoniae; M129-B7
Catalog No. 502382435
Encompass
Mycoplasma pneumoniae strain M129-B7 was isolated from a patient with pneumonia. This whole-genome sequenced bacterium has applications in infectious and respiratory disease research and it is the virulent parent strain of Mycoplasma pneumoniae strain M129-B170 (ATCC 29343).
Catalog No. 50-238-2435 Supplier ATCC Supplier No. 29342
Mycoplasma pneumoniae strain M129-B7 was isolated from a patient with pneumonia. This whole-genome sequenced bacterium has applications in infectious and respiratory disease research and it is the virulent parent strain of Mycoplasma pneumoniae strain M129-B170 (ATCC 29343).