Learn More
ATCC Mycoplasma pneumoniae; Quantitative CRM; genomic DNA
Catalog No. 502384564
Encompass
Quantified genomic DNA isolated from Mycoplasma pneumoniae FH strain of Eaton Agent [NCTC 10119] (ATCC 15531). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-4564 Supplier ATCC Supplier No. QCRM15531D
Quantified genomic DNA isolated from Mycoplasma pneumoniae FH strain of Eaton Agent [NCTC 10119] (ATCC 15531). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.