Learn More
ATCC Mycobacterium avium; subsp. silvaticum
Catalog No. 502380448
Encompass
Mycobacterium avium subsp. silvaticum strain 6409 is a bacterial type strain that was isolated from the liver and spleen of a wood pigeon in France.
Catalog No. 50-238-0448 Supplier ATCC Supplier No. 49884
Mycobacterium avium subsp. silvaticum strain 6409 is a bacterial type strain that was isolated from the liver and spleen of a wood pigeon in France.