Learn More
Bioss Mycoplasma pnuemoniae
SDP
Catalog No. 5000065309
Encompass_Preferred
Mycoplasma pnuemoniae Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65309 Supplier Bioss Supplier No. BSM72335M
Mycoplasma pnuemoniae Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide