Learn More
ATCC Mycoplasma pulmonis
Catalog No. 502382817
Encompass
Mycoplasma pulmonis strain [Ash, PG34; L3] is a bacterial type strain that was isolated from the lung lesion of a rat. Growth of this strain is best achieved if broth cultures are allowed to incubate aerobically and the plates in 5% CO2.
Catalog No. 50-238-2817 Supplier ATCC Supplier No. 19612
Mycoplasma pulmonis strain [Ash, PG34; L3] is a bacterial type strain that was isolated from the lung lesion of a rat. Growth of this strain is best achieved if broth cultures are allowed to incubate aerobically and the plates in 5% CO2.