Learn More
ATCC Halorubrum sodomense
Supplier:  ATCC 33755
Encompass
Halorubrum sodomense strain RD-26 is a halophilic archaeon that was isolated from the Dead Sea. This strain is cited to produce bacteriorhodopsin, glucoamylase, and thermophilic extracellular amyloglucosidase.
Catalog No. 50-237-9544