Learn More
ATCC Mycoplasma salivarium; Quantitative CRM; genomic DNA
Supplier:  ATCC QCRM23064D
Encompass
Quantified genomic DNA isolated from Mycoplasma salivarium strain [H110, NCTC 10113, PG 20] (ATCC 23064). This quantitative external control was produced under an ISO 17034 accredited process for use in testing and calibration in ISO 17025 accredited laboratories, inclusivity/exclusivity testing, establishing limits of detection, verification or comparison of test methods, and other molecular applications.
Catalog No. 50-238-3260
 Restricted Item