Learn More
ATCC Mycoplasma synoviae
Catalog No. 502383974
Encompass
Mycoplasma synoviae strain WVU 1853 [NCTC 10124] is a whole-genome sequenced type strain that was isolated from the hock joint of a chicken. This bacterial culture has applications in agricultural research, media testing, quality control, and pharmaceutical testing.
Catalog No. 50-238-3974 Supplier ATCC Supplier No. 25204
Add to Cart
Add to Cart
Mycoplasma synoviae strain WVU 1853 [NCTC 10124] is a whole-genome sequenced type strain that was isolated from the hock joint of a chicken. This bacterial culture has applications in agricultural research, media testing, quality control, and pharmaceutical testing.