Learn More
Romer Labs MycoSep 224 AflaZon, clean-up column, 25/pkg, push-through format, 3-5 mL volume of extract, single-use, 1-step clean-up method
Catalog No. NC1428574
Encompass
MycoSep 224 AflaZon, clean-up column, 25/pkg, push-through format, 3-5 mL volume of extract, single-use, 1-step clean-up method
Catalog No. NC1428574 Supplier Romer Labs Supplier No. 10001951
MycoSep 224 AflaZon, clean-up column, 25/pkg, push-through format, 3-5 mL volume of extract, single-use, 1-step clean-up method