Learn More
Invivogen MycoStrip™ 100 Mycoplasma Detection Kit, Rapid Detection In Under 1 Hour, High Sensitivity Detection Down To 10 CFU/ml - 100 Tests
SDP
Catalog No. NC2683201
Encompass
MycoStrip™ is a rapid and sensitive mycoplasma detection kit that allows researchers to identify contamination in cell cultures within an hour. With results available in 2-5 minutes and requiring minimal hands-on time, this kit simplifies routine testing. Its specificity for common mycoplasma species and lack of cross-reactivity set it apart from other options.
Catalog No. NC2683201 Supplier Invivogen Supplier No. REPMYSNC100
MycoStrip™ is a rapid and sensitive mycoplasma detection kit that allows researchers to identify contamination in cell cultures within an hour. With results available in 2-5 minutes and requiring minimal hands-on time, this kit simplifies routine testing. Its specificity for common mycoplasma species and lack of cross-reactivity set it apart from other options.