Learn More
Lonza Walkersville MycoZap Prophylactic
Catalog No. NC9023832
Encompass
MycoZap Prophylactic is an extremely powerful combination of innovative antibiotics for the protection of cell cultures against mycoplasma contamination. MycoZap Prophylactic specifically prevents mycoplasma contamination from growth and can be combined with other antibiotic formula of choice.Benefits:Efficient mollicute elimination by a combination of antibiotic and antimetabolic agentsMinimal toxic effectsApplications:Can be used in combination with other antibiotics (e.g., Pen/Strep) to prevent other microbial contaminantsContent:10 x 1 mL MycoZap Prophylactic
Catalog No. NC9023832 Supplier Lonza Walkersville Supplier No. VZA2031
MycoZap Prophylactic is an extremely powerful combination of innovative antibiotics for the protection of cell cultures against mycoplasma contamination. MycoZap Prophylactic specifically prevents mycoplasma contamination from growth and can be combined with other antibiotic formula of choice.Benefits:Efficient mollicute elimination by a combination of antibiotic and antimetabolic agentsMinimal toxic effectsApplications:Can be used in combination with other antibiotics (e.g., Pen/Strep) to prevent other microbial contaminantsContent:10 x 1 mL MycoZap Prophylactic