Learn More
Medchemexpress LLC HY-100669 2mg Medchemexpress, Mycro 3 CAS:944547-46-0 Purity:>98%
SDP
Catalog No. 501931911
Encompass_Preferred
Medchemexpress, HY-100669 2mg Mycro 3 CAS:944547-46-0 Mycro 3 is a potent and selective c-Myc inhibitor in whole cell assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1911 Supplier Medchemexpress LLC Supplier No. HY1006692MG
Medchemexpress, HY-100669 2mg Mycro 3 CAS:944547-46-0 Mycro 3 is a potent and selective c-Myc inhibitor in whole cell assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.