Learn More
Medchemexpress LLC HY-100669 5mg , Mycro 3 CAS:944547-46-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006695MG
Encompass_Preferred
HY-100669 5mg Mycro 3 CAS: 944547-46-0 Mycro 3 is a potent and selective inhibitor of Myc-associated factor X (MAX) dimerization. Mycro 3 also inhibit DNA binding of c-Myc. Mycro 3 could be used for the research of pancreatic cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9867