Learn More
Cell Signaling Technology Myddosome Complex Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502726626
Encompass
The Myddosome Complex Antibody Sampler Kit provides an economical means of detecting components of the myddosome complex, a large oligomeric signaling organelle formed during TLR signaling. This kit includes phospho-specific and control antibodies to facilitate the analysis of key myddosome components, including MyD88, IRAKs, TRAF6, and TBK1, via western blotting and other validated applications.
  • Contains enough antibodies to perform two western blot experiments with each primary antibody
  • Includes validated primary antibodies against key myddosome signaling proteins
  • Supports detection of both total and phospho-specific protein levels
  • Suitable for western blotting, immunoprecipitation, immunofluorescence, flow cytometry, and fluorescent western analysis
  • Supplied in a convenient kit format for efficient pathway profiling
Catalog No. 50-272-6626 Supplier Cell Signaling Technology Supplier No. 39433T
The Myddosome Complex Antibody Sampler Kit provides an economical means of detecting components of the myddosome complex, a large oligomeric signaling organelle formed during TLR signaling. This kit includes phospho-specific and control antibodies to facilitate the analysis of key myddosome components, including MyD88, IRAKs, TRAF6, and TBK1, via western blotting and other validated applications.
  • Contains enough antibodies to perform two western blot experiments with each primary antibody
  • Includes validated primary antibodies against key myddosome signaling proteins
  • Supports detection of both total and phospho-specific protein levels
  • Suitable for western blotting, immunoprecipitation, immunofluorescence, flow cytometry, and fluorescent western analysis
  • Supplied in a convenient kit format for efficient pathway profiling